COM448668274 Combi-Blocks, Inc.

2,2-Dimethylpentanenitrile

  • MDL number MFCD00055630
  • Cas Number : 20654-47-1
  • Molecular Weight : 111.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5166

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5166
Availability details for
Quantity
  • Availability
  • Add to Cart