COM448668275 Combi-Blocks, Inc.

4-Dimethylamino-4'-nitrosodiphenylamine

  • MDL number MFCD02684074
  • Cas Number : 7696-70-0
  • Molecular Weight : 241.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5188

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5188
Availability details for
Quantity
  • Availability
  • Add to Cart