COM448668277 Combi-Blocks, Inc.

2,5-Dibromothieno[2,3-b]thiophene

  • MDL number MFCD18451757
  • Cas Number : 25121-86-2
  • Molecular Weight : 298.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5213

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5213
Availability details for
Quantity
  • Availability
  • Add to Cart