COM448668281 Combi-Blocks, Inc.

2,3,5,6-Tetrafluoro-4-mercaptobenzoic acid

  • MDL number MFCD00129954
  • Cas Number : 5211-44-9
  • Molecular Weight : 226.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5292

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5292
Availability details for
Quantity
  • Availability
  • Add to Cart