COM448668284 Combi-Blocks, Inc.

2-Methyl-2-adamantyl methacrylate

  • MDL number MFCD04114354
  • Cas Number : 177080-67-0
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5307

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5307
Availability details for
Quantity
  • Availability
  • Add to Cart