COM448668299 Combi-Blocks, Inc.

(S)-4-Benzyloxy-1,3-butanediol

  • MDL number MFCD11846118
  • Cas Number : 85418-23-1
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5691

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5691
Availability details for
Quantity
  • Availability
  • Add to Cart