COM448668300 Combi-Blocks, Inc.

1-(2-Aminoethyl)-4-methylpiperazine HCl

  • MDL number MFCD20483879
  • Cas Number : 401817-30-9
  • Molecular Weight : 179.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5694

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5694
Availability details for
Quantity
  • Availability
  • Add to Cart