COM448668303 Combi-Blocks, Inc.

O-Acetyl-n-methylhydroxylamine HCl

  • MDL number MFCD22581435
  • Cas Number : 19689-95-3
  • Molecular Weight : 125.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5717

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5717
Availability details for
Quantity
  • Availability
  • Add to Cart