COM448668307 Combi-Blocks, Inc.

2-Ethyl-2-adamantyl methacrylate

  • MDL number MFCD06411144
  • Cas Number : 209982-56-9
  • Molecular Weight : 248.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5850

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5850
Availability details for
Quantity
  • Availability
  • Add to Cart