COM448668311 Combi-Blocks, Inc.

1,3-Bis(3-aminophenoxy)-5-chlorobenzene diHCl

  • MDL number MFCD14708170
  • Cas Number : 99503-73-8
  • Molecular Weight : 399.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5993

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-5993
Availability details for
Quantity
  • Availability
  • Add to Cart