COM448668323 Combi-Blocks, Inc.

(S)-4-(4-Methoxybenzyloxy)-1,3-butanediol

  • MDL number MFCD11870115
  • Cas Number : 1820579-68-7
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6142

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6142
Availability details for
Quantity
  • Availability
  • Add to Cart