COM448668326 Combi-Blocks, Inc.

4-(2-Chloroethylsulfonyl)butanoic acid

  • MDL number MFCD13185961
  • Cas Number : 121315-24-0
  • Molecular Weight : 214.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6155

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6155
Availability details for
Quantity
  • Availability
  • Add to Cart