COM448668327 Combi-Blocks, Inc.

2-Methoxy-1-naphthalenemethanol

  • MDL number MFCD00274224
  • Cas Number : 40696-22-8
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6172

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6172
Availability details for
Quantity
  • Availability
  • Add to Cart