COM448668330 Combi-Blocks, Inc.

Dimethyl 1,8-anthracenedicarboxylate

  • MDL number MFCD01321126
  • Cas Number : 93655-34-6
  • Molecular Weight : 294.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6228

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6228
Availability details for
Quantity
  • Availability
  • Add to Cart