COM448668334 Combi-Blocks, Inc.

Dodecyl ether

  • MDL number MFCD00026590
  • Cas Number : 4542-57-8
  • Molecular Weight : 354.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6371

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6371
Availability details for
Quantity
  • Availability
  • Add to Cart