COM448668338 Combi-Blocks, Inc.

4-Methyl-2-pyridyl trifluoromethanesulfonate

  • MDL number MFCD11042681
  • Cas Number : 179260-78-7
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6450

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6450
Availability details for
Quantity
  • Availability
  • Add to Cart