COM448668340 Combi-Blocks, Inc.

Tributyl(2-methoxyethyl)phosphonium bis(trifluoromethanesulfonyl)imide

  • MDL number MFCD11112190
  • Cas Number : 959698-44-3
  • Molecular Weight : 541.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6470

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6470
Availability details for
Quantity
  • Availability
  • Add to Cart