COM448668342 Combi-Blocks, Inc.

2,5-Di(2-thienyl)-1h-pyrrole

  • MDL number MFCD03931087
  • Cas Number : 89814-62-0
  • Molecular Weight : 231.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6513

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6513
Availability details for
Quantity
  • Availability
  • Add to Cart