COM448668346 Combi-Blocks, Inc.

2-Bromo-7-tert-butylpyrene

  • MDL number MFCD23143151
  • Cas Number : 78751-80-1
  • Molecular Weight : 337.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6566

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6566
Availability details for
Quantity
  • Availability
  • Add to Cart