COM448668350 Combi-Blocks, Inc.

Chloromethyl 2,2,2-trichloroethyl ether

  • MDL number MFCD20257906
  • Cas Number : 69573-75-7
  • Molecular Weight : 197.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6715

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6715
Availability details for
Quantity
  • Availability
  • Add to Cart