COM448668352 Combi-Blocks, Inc.

Isobutylcyclohexane

  • MDL number MFCD00013767
  • Cas Number : 1678-98-4
  • Molecular Weight : 140.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6791

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-6791
Availability details for
Quantity
  • Availability
  • Add to Cart