COM448668361 Combi-Blocks, Inc.

2,5-Dihydroxyphenyl(diphenyl)phosphine oxide

  • MDL number MFCD00408436
  • Cas Number : 13291-46-8
  • Molecular Weight : 310.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7055

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7055
Availability details for
Quantity
  • Availability
  • Add to Cart