COM448668364 Combi-Blocks, Inc.

1,3-Bis[(2,2-dimethyl-1,3-dioxan-5-yl)oxy]-2-propanol

  • MDL number MFCD22041644
  • Cas Number : 847682-00-2
  • Molecular Weight : 320.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7091

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7091
Availability details for
Quantity
  • Availability
  • Add to Cart