COM448668367 Combi-Blocks, Inc.

(4S,4'S)-2,2'-(Pentane-3,3'-diyl)bis(4-benzyl-4,5-dihydrooxazole)

  • MDL number MFCD20265601
  • Cas Number : 160191-64-0
  • Molecular Weight : 390.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7136

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7136
Availability details for
Quantity
  • Availability
  • Add to Cart