COM448668369 Combi-Blocks, Inc.

9-(P-Tolyl)carbazole

  • MDL number MFCD14708175
  • Cas Number : 19264-73-4
  • Molecular Weight : 257.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7176

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7176
Availability details for
Quantity
  • Availability
  • Add to Cart