COM448668374 Combi-Blocks, Inc.

1-(2-Amino-1-naphthyl)isoquinoline

  • MDL number MFCD12407125
  • Cas Number : 664302-70-9
  • Molecular Weight : 270.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7327

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7327
Availability details for
Quantity
  • Availability
  • Add to Cart