COM448668378 Combi-Blocks, Inc.

6-Methoxypurine

  • MDL number MFCD00005574
  • Cas Number : 1074-89-1
  • Molecular Weight : 150.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7598
Availability details for
Quantity
  • Availability
  • Add to Cart