COM448668385 Combi-Blocks, Inc.

1-Methyl-4-(1,4'-bipiperidin-4-yl)piperazine

  • MDL number MFCD20257913
  • Cas Number : 1629218-99-0
  • Molecular Weight : 266.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7797

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7797
Availability details for
Quantity
  • Availability
  • Add to Cart