COM448668389 Combi-Blocks, Inc.

1-Butyl-3-methylimidazolium trifluoro(trifluoromethyl)borate

  • MDL number MFCD15072148
  • Cas Number : 741677-68-9
  • Molecular Weight : 276.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7959

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-7959
Availability details for
Quantity
  • Availability
  • Add to Cart