COM448668395 Combi-Blocks, Inc.

1-Hexyl-2,3-dimethylimidazolium iodide

  • MDL number MFCD18207720
  • Cas Number : 288627-94-1
  • Molecular Weight : 308.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8084

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8084
Availability details for
Quantity
  • Availability
  • Add to Cart