COM448668397 Combi-Blocks, Inc.

(R)-Benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5h-pyrrolo[2,1-c][1,2,4]triazolium tetrafluoroborate

  • MDL number MFCD23380201
  • Cas Number : 862095-77-0
  • Molecular Weight : 431.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8103

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8103
Availability details for
Quantity
  • Availability
  • Add to Cart