COM448668401 Combi-Blocks, Inc.

4-Methoxyphenyl 2-o-acetyl-3-o-allyl-4,6-o-benzylidene-beta-d-glucopyranoside

  • MDL number MFCD20265596
  • Cas Number : 1477956-18-5
  • Molecular Weight : 456.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8147

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8147
Availability details for
Quantity
  • Availability
  • Add to Cart