COM448668403 Combi-Blocks, Inc.

2-(1,3-Benzodioxol-5-yl)-4,6-bis(trichloromethyl)-1,3,5-triazine

  • MDL number MFCD04038074
  • Cas Number : 71255-78-2
  • Molecular Weight : 435.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8301

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8301
Availability details for
Quantity
  • Availability
  • Add to Cart