COM448668422 Combi-Blocks, Inc.

Sodium 2,3,5,6-tetrafluoro-4-hydroxybenzenesulfonate

  • MDL number MFCD03427812
  • Cas Number : 221908-17-4
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8687

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8687
Availability details for
Quantity
  • Availability
  • Add to Cart