COM448668423 Combi-Blocks, Inc.

2-Bromo-9,10-diphenylanthracene

  • MDL number MFCD14708167
  • Cas Number : 201731-79-5
  • Molecular Weight : 409.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8707

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8707
Availability details for
Quantity
  • Availability
  • Add to Cart