COM448668437 Combi-Blocks, Inc.

Bismuth sodium ethylenediaminetetraacetate

  • MDL number MFCD00070494
  • Cas Number : 12558-49-5
  • Molecular Weight : 520.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8951

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-8951
Availability details for
Quantity
  • Availability
  • Add to Cart