COM448668442 Combi-Blocks, Inc.

1-Methyl-3-[6-(methylthio)hexyl]imidazolium p-toluenesulfonate

  • MDL number MFCD23380210
  • Cas Number : 1352947-63-7
  • Molecular Weight : 384.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9145

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9145
Availability details for
Quantity
  • Availability
  • Add to Cart