COM448668444 Combi-Blocks, Inc.

Bis[2-(trimethylsilyloxy)ethyl] ether

  • MDL number MFCD11975427
  • Cas Number : 16654-74-3
  • Molecular Weight : 250.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9204

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9204
Availability details for
Quantity
  • Availability
  • Add to Cart