COM448668452 Combi-Blocks, Inc.

2,2',7,7'-Tetrabromo-9,9'-bifluorenylidene

  • MDL number MFCD22581445
  • Cas Number : 27192-91-2
  • Molecular Weight : 644.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9326
Availability details for
Quantity
  • Availability
  • Add to Cart