COM448668456 Combi-Blocks, Inc.

2,3-Epoxynorbornane

  • MDL number MFCD00798067
  • Cas Number : 278-74-0
  • Molecular Weight : 110.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9404

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9404
Availability details for
Quantity
  • Availability
  • Add to Cart