COM448668457 Combi-Blocks, Inc.

5-Norbornene-2-carboxamide

  • MDL number MFCD00187896
  • Cas Number : 95-17-0
  • Molecular Weight : 137.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9424

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9424
Availability details for
Quantity
  • Availability
  • Add to Cart