COM448668458 Combi-Blocks, Inc.

Diphenyl 4-bromo-alpha-chlorobenzylphosphonate

  • MDL number MFCD15072158
  • Cas Number : 189099-56-7
  • Molecular Weight : 437.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9425

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9425
Availability details for
Quantity
  • Availability
  • Add to Cart