COM448668461 Combi-Blocks, Inc.

Diphenyl-1-pyrenylphosphine

  • MDL number MFCD00278755
  • Cas Number : 110231-30-6
  • Molecular Weight : 386.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9461

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9461
Availability details for
Quantity
  • Availability
  • Add to Cart