COM448668463 Combi-Blocks, Inc.

Amyltriethylammonium bis(trifluoromethanesulfonyl)imide

  • MDL number MFCD15072143
  • Cas Number : 906478-91-9
  • Molecular Weight : 452.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9554

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9554
Availability details for
Quantity
  • Availability
  • Add to Cart