COM448668473 Combi-Blocks, Inc.

3,4'-Diamino-4-nitrodiphenyl ether

  • MDL number MFCD15072156
  • Cas Number : 30491-74-8
  • Molecular Weight : 245.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9739

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9739
Availability details for
Quantity
  • Availability
  • Add to Cart