COM448668479 Combi-Blocks, Inc.

Diallyl 1,4-cyclohexanedicarboxylate

  • MDL number MFCD00050452
  • Cas Number : 20306-22-3
  • Molecular Weight : 252.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9849

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9849
Availability details for
Quantity
  • Availability
  • Add to Cart