COM448668481 Combi-Blocks, Inc.

1-Ethyl-2,3-dimethylimidazolium bis(trifluoromethanesulfonyl)imide

  • MDL number MFCD10566900
  • Cas Number : 174899-90-2
  • Molecular Weight : 405.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9951

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9951
Availability details for
Quantity
  • Availability
  • Add to Cart