COM448668482 Combi-Blocks, Inc.

2-Mercapto-5-phenylbenzoxazole

  • MDL number MFCD00022847
  • Cas Number : 17371-99-2
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9979

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9979
Availability details for
Quantity
  • Availability
  • Add to Cart