COM448668483 Combi-Blocks, Inc.

3,3'-Dihexyl-2,2'-bithiophene

  • MDL number MFCD23380206
  • Cas Number : 125607-30-9
  • Molecular Weight : 334.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9990

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-9990
Availability details for
Quantity
  • Availability
  • Add to Cart