COM448668532 Combi-Blocks, Inc.

4-(Methylcarbamoyl)benzoic acid

  • MDL number MFCD10688269
  • Cas Number : 23754-45-2
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4068

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4068
Availability details for
Quantity
  • Availability
  • Add to Cart